2-Cyano-5-pyrimidineboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-Cyanopyrimidine-5-boronic Acid Pinacol Ester

A618516-1G A618516-250MG A618516-5G
76980-598EA 50.93 USD
76980-598 76980-600 76980-602
2-Cyano-5-pyrimidineboronic acid pinacol ester 95%
2-Cyano-5-pyrimidineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₄BN₃O₂
MW: 231.0588 g/mol
MDL Number: MFCD08669562
CAS Number: 1025708-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR