Ethyl-4-aminopyrimidine-5-carboxylate 97%

Supplier: Ambeed
Warning

A118296-1G A118296-250MG A118296-5G
76971-344EA 30 USD
76971-344 76971-346 77880-956
Ethyl-4-aminopyrimidine-5-carboxylate 97%
Ethyl-4-aminopyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉N₃O₂
MDL Number: MFCD01028678
CAS Number: 65195-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR