3-Methoxy-5-(trifluoromethyl)benzonitrile 98%

Supplier: Ambeed
Warning

A544316-1G A544316-25G A544316-5G
76980-202EA 10.61 USD
76980-202 76980-204 76980-206
3-Methoxy-5-(trifluoromethyl)benzonitrile 98%
3-Methoxy-5-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₉H₆F₃NO
MW: 201.1453 g/mol
MDL Number: MFCD08458145
CAS Number: 868167-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR