3-Bromo-4-isopropylbenzoic acid 96%

Supplier: Ambeed
Warning

A601051-100MG A601051-1G A601051-250MG A601051-5G
76980-500EA 31.83 USD
76980-500 76980-502 76980-504 76980-506
3-Bromo-4-isopropylbenzoic acid 96%
3-Bromo-4-isopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.09 g/mol
MDL Number: MFCD11110860
CAS Number: 99070-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR