N-Methyl-2,2,2-trifluoroacetamide 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Methyltrifluoroacetamide, 2,2,2-Trifluoro-N-methylacetamide
A234459-10G
A234459-25G
A234459-500G
A234459-100G
76976-276EA
11.67
USD
76976-276
76976-278
76976-280
77672-090
N-Methyl-2,2,2-trifluoroacetamide 95%
N-Methyl-2,2,2-trifluoroacetamide
Precaución: for Research Use Only
Formula:
C₃H₄F₃NO MW: 127.07 g/mol |
MDL Number:
MFCD00009670 CAS Number: 815-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...