Ala-OBzl TosOH 98%

Supplier: Ambeed
Warning

Synonyms: L-Alanine Benzyl Ester p-Toluenesulphonate, L-Alanine benzyl ester p-toluenesulphonate salt, salt H-Ala-OBzl.Tos-OH, H-Ala-OBzl·p-tosylate, H-Ala-OBzl.TosOH, H-Ala-OBzl·Tos-OH

A682502-100G A682502-1G A682502-5G A682502-25G
76981-006EA 35 USD
76981-006 76981-008 76981-010 76981-012 77673-860
Ala-OBzl TosOH 98%
Ala-OBzl TosOH

Precaución: for Research Use Only

Formula: C₁₇H₂₁NO₅S
MDL Number: MFCD00066143
CAS Number: 42854-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR