4-Spiroindanepiperidine hydrochloride 98%

Supplier: Ambeed
Warning

A521460-100MG A521460-1G A521460-250MG A521460-5G
76980-022EA 14 USD
76980-022 76980-024 76980-026 77673-386
4-Spiroindanepiperidine hydrochloride 98%
4-Spiroindanepiperidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₈ClN
MDL Number: MFCD08461061
CAS Number: 96651-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR