2-Amino-4-(trifluoromethyl)thiophenol hydrochloride 95%

Supplier: Ambeed
Danger

A620613-1G A620613-5G A620613-25G
76980-616EA 26 USD
76980-616 76980-618 77882-448
2-Amino-4-(trifluoromethyl)thiophenol hydrochloride 95%
2-Amino-4-(trifluoromethyl)thiophenol hydrochloride

Precaución: for Research Use Only

Formula: C₇H₇ClF₃NS
MDL Number: MFCD00042600
CAS Number: 4274-38-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR