Cbz-N-methyl-L-valine 97%

Supplier: Ambeed
Warning

A514630-10G A514630-1G A514630-250MG A514630-25G A514630-5G A514630-100G
76979-956EA 79 USD
76979-956 76979-958 76979-960 76979-962 76979-964 77673-366
Cbz-N-methyl-L-valine 97%
Cbz-N-methyl-L-valine

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
MDL Number: MFCD00153397
CAS Number: 42417-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR