3-Amino-1-methylpiperidine dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methylpiperidin-3-amine dihydrochloride

A281255-1G A281255-250MG A281255-25G A281255-5G
76977-406EA 15 USD
76977-406 76977-408 76977-410 76977-412
3-Amino-1-methylpiperidine dihydrochloride 98%
3-Amino-1-methylpiperidine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₆Cl₂N₂
MW: 187.11 g/mol
MDL Number: MFCD06796280
CAS Number: 894808-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR