(S)-2-(Methylamino)succinic acid 95%

Supplier: Ambeed
Warning

Synonyms: N-Me-Asp-OH, N-Methyl-L-aspartic acid

A137010-100MG A137010-1G A137010-250MG
76972-376EA 25 USD
76972-376 76972-378 76972-380
(S)-2-(Methylamino)succinic acid 95%
(S)-2-(Methylamino)succinic acid

Precaución: for Research Use Only

Formula: C₅H₉NO₄
MDL Number: MFCD00069561
CAS Number: 4226-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR