5-Ethyluracil 98%

Supplier: Ambeed
Warning

Synonyms: Ethyluracil, 5-Ethylpyrimidine-2,4(1H,3H)-dione

A273638-100G A273638-10G A273638-1G A273638-25G A273638-5G
76977-230EA 35.01 USD
76977-230 76977-232 76977-234 76977-236 76977-238
5-Ethyluracil 98%
5-Ethyluracil

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
MDL Number: MFCD00079187
CAS Number: 4212-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR