1-(4-Bromophenyl)-2-chloroethanone 98%

Supplier: Ambeed
Danger

A125362-100G A125362-10G A125362-1G A125362-25G A125362-5G
76971-816EA 118.01 USD
76971-816 76971-818 76971-820 76971-822 76971-824
1-(4-Bromophenyl)-2-chloroethanone 98%
1-(4-Bromophenyl)-2-chloroethanone

Precaución: for Research Use Only

Formula: C₈H₆BrClO
MDL Number: MFCD00095178
CAS Number: 4209-02-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR