6-Propylpyridin-2-amine 95%

Supplier: Ambeed
Danger

A431566-1G A431566-250MG
76979-304EA 287 USD
76979-304 76979-306
6-Propylpyridin-2-amine 95%
6-Propylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂
MDL Number: MFCD00130076
CAS Number: 41995-29-3
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR