Methyl-2-piperidinecarboxylate 98%

Supplier: Ambeed
Warning

A469052-10G A469052-25G A469052-5G A469052-100G
76979-622EA 6 USD
76979-622 76979-624 76979-626 77882-190
Methyl-2-piperidinecarboxylate 98%
Methyl 2-piperidinecarboxylate

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MDL Number: MFCD07364347
CAS Number: 41994-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR