3-Amino-5-bromopyridin-4-ol 97%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-5-bromo-4-hydroxypyridine

A104376-100MG A104376-1G A104376-250MG A104376-5G
76970-334EA 12 USD
76970-334 76970-336 76970-338 76970-340
3-Amino-5-bromopyridin-4-ol 97%
3-Amino-5-bromopyridin-4-ol

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O
MW: 189.012 g/mol
MDL Number: MFCD08692340
CAS Number: 101084-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR