Penta-O-acetyl-β-D-galactopyranose 98%

Supplier: Ambeed
Warning

Synonyms: β-D-Galactose Pentaacetate, (2S,3R,4S,5S,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate

A335850-100G A335850-10G A335850-25G A335850-500G A335850-5G A335850-1000G
76978-348EA 29 USD
76978-348 76978-350 76978-352 76978-354 76978-356 77881-929
Penta-O-acetyl-β-D-galactopyranose 98%
Penta-O-acetyl-beta-D-galactopyranose

Precaución: for Research Use Only

Formula: C₁₆H₂₂O₁₁
MDL Number: MFCD00063259
CAS Number: 4163-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR