2-(2-Pyridinyl)-1H-benzimidazole-6-carboxylic acid 95%

Supplier: Ambeed
Warning

A197583-100MG A197583-250MG
76975-108EA 100 USD
76975-108 76975-110
2-(2-Pyridinyl)-1H-benzimidazole-6-carboxylic acid 95%
2-(2-Pyridinyl)-1H-benzimidazole-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₉N₃O₂
MDL Number: MFCD06245483
CAS Number: 669070-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR