4-Cyano-4'-heptylbiphenyl 98%

Supplier: Ambeed
Warning

A137261-1G A137261-250MG A137261-25G A137261-5G A137261-10G
76972-390EA 9 USD
76972-390 76972-392 76972-394 76972-396 77881-081
4-Cyano-4'-heptylbiphenyl 98%
4-Cyano-4'-heptylbiphenyl

Precaución: for Research Use Only

Formula: C₂₀H₂₃N
MDL Number: MFCD00074915
CAS Number: 41122-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR