Methyl-4-acetamido-5-chloro-2-methoxybenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-acetamido-5-chloro-2-methoxybenzoate, Metoclopramide Related Compound B
A359003-100G
A359003-10G
A359003-25G
A359003-5G
A359003-500G
76978-566EA
68
USD
76978-566
76978-568
76978-570
76978-572
77672-804
Methyl-4-acetamido-5-chloro-2-methoxybenzoate 98%
Methyl-4-acetamido-5-chloro-2-methoxybenzoate
Precaución: for Research Use Only
Formula:
C₁₁H₁₂ClNO₄ |
MDL Number:
MFCD00048191 CAS Number: 4093-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...