2-Amino-4-bromophenol 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-hydroxyaniline

A463737-10G A463737-1G A463737-25G A463737-5G
76979-576EA 15.92 USD
76979-576 76979-578 76979-580 76979-582
2-Amino-4-bromophenol 98%
2-Amino-4-bromophenol

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.02 g/mol
MDL Number: MFCD00235171
CAS Number: 40925-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR