2,5-Dimethyl-4-methoxy-3(2H)-furanone 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxy-2,5-dimethylfuran-3(2H)-one, 4-Methoxy-2,5-dimethyl-3(2H)-furanone

A282395-100G A282395-25G A282395-1G
76977-458EA 147.47 USD
76977-458 76977-460 77672-460
2,5-Dimethyl-4-methoxy-3(2H)-furanone 97%
2,5-Dimethyl-4-methoxy-3(2H)-furanone

Precaución: for Research Use Only

Formula: C₇H₁₀O₃
MW: 142.15 g/mol
MDL Number: MFCD00209504
CAS Number: 4077-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR