1-Bromo-4-(trifluoromethoxy)benzene 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Trifluoromethoxy)bromobenzene

A229077-1000G A229077-100G A229077-10G A229077-1G A229077-25G A229077-500G A229077-5G
76976-072EA 140.04 USD
76976-072 76976-074 76976-076 76976-078 76976-080 76976-082 76976-084
1-Bromo-4-(trifluoromethoxy)benzene 98%
1-Bromo-4-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₄BrF₃O
MW: 241.01 g/mol
MDL Number: MFCD00040834
CAS Number: 407-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR