2-Amino-5-methylbenzamide 97%

Supplier: Ambeed
Warning

A103830-1G A103830-250MG A103830-5G
76970-304EA 45 USD
76970-304 76970-306 76970-308
2-Amino-5-methylbenzamide 97%
2-Amino-5-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O
MDL Number: MFCD03428521
CAS Number: 40545-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR