1-Benzhydrylazetidin-3-amine 97%

Supplier: Ambeed
Warning

A315266-10G A315266-1G A315266-25G A315266-5G
76977-972EA 37 USD
76977-972 76977-974 76977-976 76977-978
1-Benzhydrylazetidin-3-amine 97%
1-Benzhydrylazetidin-3-amine

Precaución: for Research Use Only

Formula: C₁₆H₁₈N₂
MDL Number: MFCD03093386
CAS Number: 40432-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR