4-Amino-1-(methylsulfonyl)piperidine 97%

Supplier: Ambeed
Warning

A116759-10G A116759-1G A116759-250MG A116759-25G A116759-5G A116759-100G
76971-242EA 32 USD
76971-242 76971-244 76971-246 76971-248 76971-250 77599-688
4-Amino-1-(methylsulfonyl)piperidine 97%
4-Amino-1-(methylsulfonyl)piperidine

Precaución: for Research Use Only

Formula: C₆H₁₄N₂O₂S
MDL Number: MFCD06805736
CAS Number: 402927-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR