5-Acetylsalicylamide 98%

Supplier: Ambeed
Warning

Synonyms: 5-Acetyl-2-hydroxybenzamide

A159415-100G A159415-10G A159415-25G
76973-470EA 49 USD
76973-470 76973-472 76973-474
5-Acetylsalicylamide 98%
5-Acetylsalicylamide

Precaución: for Research Use Only

Formula: C₉H₉NO₃
MW: 179.18 g/mol
MDL Number: MFCD00049222
CAS Number: 40187-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR