2-Amino-3-chloro-5-nitrobenzotrifluoride 97%

Supplier: Ambeed
Warning

A151357-250MG A151357-5G
76973-120EA 11.67 USD
76973-120 76973-122
2-Amino-3-chloro-5-nitrobenzotrifluoride 97%
2-Amino-3-chloro-5-nitrobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₄ClF₃N₂O₂
MW: 240.57 g/mol
MDL Number: MFCD04972736
CAS Number: 400-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR