2-[(5-Bromo-4-methyl-2-pyridinyl)amino]-1-ethanol 95%

Supplier: Ambeed
Warning

Synonyms: 2-((5-Bromo-4-methylpyridin-2-yl)amino)ethanol

A541763-100MG A541763-1G A541763-250MG
76980-190EA 17 USD
76980-190 77882-314 77882-315
2-[(5-Bromo-4-methyl-2-pyridinyl)amino]-1-ethanol 95%
2-[(5-Bromo-4-methyl-2-pyridinyl)amino]-1-ethanol

Precaución: for Research Use Only

Formula: C₈H₁₁BrN₂O
MDL Number: MFCD13562416
CAS Number: 1219982-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR