1-Fluoro-2-(2-nitrovinyl)benzene 95%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-beta-nitrostyrene, 1-Fluoro-2-(2-nitroethenyl)benzene

A122550-1G A122550-250MG A122550-5G
76971-628EA 33.95 USD
76971-628 76971-630 76971-632
1-Fluoro-2-(2-nitrovinyl)benzene 95%
1-Fluoro-2-(2-nitrovinyl)benzene

Precaución: for Research Use Only

Formula: C₈H₆FNO₂
MW: 167.14 g/mol
MDL Number: MFCD00042451
CAS Number: 399-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR