2-Amino-5-bromobenzophenone 97%

Supplier: Ambeed
Warning

A446736-250MG A446736-25G A446736-5G A446736-1G A446736-100G
76979-440EA 12.74 USD
76979-440 76979-442 76979-444 76979-446 77673-098 77882-156
2-Amino-5-bromobenzophenone 97%
2-Amino-5-bromobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrNO
MDL Number: MFCD00059962
CAS Number: 39859-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR