(S)-Isopropyl-2-aminopropanoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (S)-Isopropyl 2-aminopropanoate hydrochloride, (S)-2-Aminopropionic acid Isopropyl ester hydrochloride, L-Alanine Isopropyl ester hydrochloride, H-Ala-OiPr·HCl, Isopropyl (S)-2-Aminopropionate hydrochloride

A169568-100G A169568-10G A169568-25G A169568-500G A169568-5G
76973-918EA 23.34 USD
76973-918 76973-920 76973-922 76973-924 76973-926
(S)-Isopropyl-2-aminopropanoate hydrochloride 97%
(S)-Isopropyl-2-aminopropanoate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂·HCl
MW: 167.6 g/mol
MDL Number: MFCD08059709
CAS Number: 62062-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR