2-Bromo-6-phenylpyridine 95%

Supplier: Ambeed
Warning

A207898-1G A207898-250MG A207898-5G
76975-412EA 34 USD
76975-412 76975-414 77881-491
2-Bromo-6-phenylpyridine 95%
2-Bromo-6-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₈BrN
MDL Number: MFCD00234691
CAS Number: 39774-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR