2-Iodo-6-methoxybenzoic acid 97%

Supplier: Ambeed
Warning

A234204-10G A234204-1G A234204-250MG A234204-25G A234204-5G
76976-260EA 203 USD
76976-260 76976-262 76976-264 76976-266 76976-268
2-Iodo-6-methoxybenzoic acid 97%
2-Iodo-6-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇IO₃
MDL Number: MFCD13562947
CAS Number: 66195-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR