Ethyl-2-((4-methoxybenzyl)amino)acetate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-((4-methoxybenzyl)amino)acetate

A144570-1G A144570-250MG A144570-5G
76972-750EA 42.44 USD
76972-750 76972-752 77671-050
Ethyl-2-((4-methoxybenzyl)amino)acetate 97%
Ethyl-2-((4-methoxybenzyl)amino)acetate

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₃
MW: 223.2700 g/mol
MDL Number: MFCD11151999
CAS Number: 60857-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR