2-Chloro-N-(4-methoxyphenyl)nicotinamide 98%

Supplier: Ambeed
Warning

A160754-100MG A160754-1G A160754-250MG
76973-580EA 9 USD
76973-580 76973-582 76973-584
2-Chloro-N-(4-methoxyphenyl)nicotinamide 98%
2-Chloro-N-(4-methoxyphenyl)nicotinamide

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClN₂O₂
MDL Number: MFCD00665154
CAS Number: 56149-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR