(4-Acetamidophenoxy)acetic acid 95+%

Supplier: Ambeed
Warning

A771629-1G A771629-250MG A771629-5G
76981-506EA 6.37 USD
76981-506 76981-508 76981-510 76981-512
(4-Acetamidophenoxy)acetic acid 95+%
(4-Acetamidophenoxy)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₄
MDL Number: MFCD01242445
CAS Number: 39149-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR