L(+)-Lysine monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: L-(+)-Lysine, Hydrate, (S)-(+)-2,6-Diaminohexanoic acid monohydrate, (S)-(+)-Lysine monohydrate, L-Lysine hydrate, (S)-(+)-2,6-Diaminocaproic acid monohydrate, H-Lys-OH monohydrate, K monohydrate, H-Lys-OH·H₂O, Lys monohydrate

A162671-100G A162671-25G A162671-500G
76973-640EA 9.55 USD
76973-640 76973-642 76973-644
L(+)-Lysine monohydrate 98%
L(+)-Lysine monohydrate

Precaución: for Research Use Only

Formula: H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
MW: 164.2 g/mol
MDL Number: MFCD00151035
CAS Number: 39665-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR