2-Amino-N-benzylacetamide hydrochloride 98%

Supplier: Ambeed
Warning

A722138-5G A722138-1G A722138-250MG
76981-240EA 167.99 USD
76981-240 77602-854 77673-980
2-Amino-N-benzylacetamide hydrochloride 98%
2-Amino-N-benzylacetamide hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₃ClN₂O
MDL Number: MFCD09909316
CAS Number: 20432-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR