4-Bromo-2-chlorophenol 96%

Supplier: Ambeed
Warning

A188042-100G A188042-5G
76974-740EA 16 USD
76974-740 76974-742 76974-744 76974-746
4-Bromo-2-chlorophenol 96%
4-Bromo-2-chlorophenol

Precaución: for Research Use Only

Formula: C₆H₄BrClO
MDL Number: MFCD00002166
CAS Number: 3964-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR