1-(2-Chloro-5-fluoropyridin-3-yl)ethanone 95%

Supplier: Ambeed
Warning

A273726-100MG A273726-1G A273726-250MG A273726-5G
76977-240EA 12 USD
76977-240 76977-242 76977-244 76977-246
1-(2-Chloro-5-fluoropyridin-3-yl)ethanone 95%
1-(2-Chloro-5-fluoropyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₅ClFNO
MDL Number: MFCD13563063
CAS Number: 1203499-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR