5-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde 97+%

Supplier: Ambeed
Warning

A220339-100MG A220339-1G A220339-250MG
76975-806EA 44.56 USD
76975-806 76975-808 76975-810
5-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde 97+%
5-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MDL Number: MFCD12962781
CAS Number: 1190321-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR