7-Bromoquinazolin-4-amine 98%

Supplier: Ambeed
Warning

A289799-100MG A289799-10G A289799-1G A289799-250MG A289799-25G A289799-5G
76977-534EA 18.04 USD
76977-534 76977-536 76977-538 76977-540 76977-542 76977-544
7-Bromoquinazolin-4-amine 98%
7-Bromoquinazolin-4-amine

Precaución: for Research Use Only

Formula: C₈H₆BrN₃
MW: 224.06 g/mol
MDL Number: MFCD09027696
CAS Number: 1123169-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR