4,6-Quinolinediol 97%

Supplier: Ambeed
Warning

Synonyms: 4,6-Dihydroxyquinoline

A148880-100MG A148880-1G A148880-250MG
76973-006EA 19.1 USD
76973-006 76973-008 76973-010
4,6-Quinolinediol 97%
4,6-Quinolinediol

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MW: 161.1574 g/mol
MDL Number: MFCD09261119
CAS Number: 3517-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR