2-Amino-5-bromopyrimidine-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A140853-1G A140853-250MG
76972-610EA 254.62 USD
76972-610 76972-612
2-Amino-5-bromopyrimidine-4-carboxylic acid 95%
2-Amino-5-bromopyrimidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₄BrN₃O₂
MW: 218.01 g/mol
MDL Number: MFCD08690775
CAS Number: 914208-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR