2-Aminopropanamide hydrochloride 95%

Supplier: Ambeed
Warning

A291539-100G A291539-1G A291539-25G A291539-5G
76977-572EA 326.76 USD
76977-572 76977-574 76977-576 76977-578
2-Aminopropanamide hydrochloride 95%
2-Aminopropanamide hydrochloride

Precaución: for Research Use Only

Formula: C₃H₉ClN₂O
MDL Number: MFCD00066144
CAS Number: 80222-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR