5-Fluoro-2-methoxybenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A292427-100G
A292427-10G
A292427-1G
A292427-250MG
A292427-25G
A292427-500G
A292427-5G
76977-596EA
293.87
USD
76977-596
76977-598
76977-600
76977-602
76977-604
76977-606
76977-608
5-Fluoro-2-methoxybenzoic acid 98%
5-Fluoro-2-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₇FO₃ MW: 170.14 g/mol |
MDL Number:
MFCD00671765 CAS Number: 394-04-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...