Ethyl cyano(hydroxyimino)acetate 95%

Supplier: Ambeed
Danger

Synonyms: Ethyl 2-cyano-2-(hydroxyimino)acetate, Ethyl Cyanoglyoxalate Oxime, Cyano(hydroxyimino)acetic Acid Ethyl Ester, Oxyma

A671796-1000G A671796-100G A671796-500G A671796-25G
76980-924EA 90.18 USD
76980-924 76980-926 76980-928 77552-046
Ethyl cyano(hydroxyimino)acetate 95%
Ethyl cyano(hydroxyimino)acetate

Precaución: for Research Use Only

Formula: C₅H₆N₂O₃
MW: 142.11 g/mol
MDL Number: MFCD00002112
CAS Number: 3849-21-6
UN: 2926
ADR: 4.1,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR