N-[3-(Dimethylamino)propyl]acrylamide 96% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A329359-25G A329359-500G A329359-100G
76978-206EA 8 USD
76978-206 76978-208 77550-924
N-[3-(Dimethylamino)propyl]acrylamide 96% stabilized
N-[3-(Dimethylamino)propyl]acrylamide

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O
MDL Number: MFCD00059902
CAS Number: 3845-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR