1-(6-Chloro-2-pyridinyl)-4-piperidinol 95%

Supplier: Ambeed
Warning

Synonyms: 1-(6-Chloropyridin-2-yl)piperidin-4-ol

A249816-1G A249816-250MG
76976-658EA 61.54 USD
76976-658 76976-660
1-(6-Chloro-2-pyridinyl)-4-piperidinol 95%
1-(6-Chloro-2-pyridinyl)-4-piperidinol

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClN₂O
MW: 212.68 g/mol
MDL Number: MFCD12923030
CAS Number: 1220036-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR